Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAAATTGATAGTTGCAGGTTAT[C/T]GCAAGATGTCTTAGAGTAGGGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602324 MIM: 612189 MIM: 610328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PBLD - phenazine biosynthesis like protein domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUFY2 - RUN and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042417.1 | 1968 | Intron | NP_001035882.1 | |||
NM_001278225.1 | 1968 | Intron | NP_001265154.1 | |||
NM_017987.4 | 1968 | Intron | NP_060457.4 | |||
XM_005269953.4 | 1968 | Intron | XP_005270010.1 | |||
XM_005269955.4 | 1968 | Intron | XP_005270012.1 | |||
XM_005269956.4 | 1968 | Intron | XP_005270013.1 | |||
XM_005269957.4 | 1968 | Intron | XP_005270014.1 | |||
XM_006717911.3 | 1968 | Intron | XP_006717974.1 | |||
XM_011539942.2 | 1968 | Intron | XP_011538244.1 |