Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAAATGACTGCAAAACCCTTTT[C/G]TTAATAATATTTTATTGTCAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT18 - polypeptide N-acetylgalactosaminyltransferase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198516.2 | 2353 | UTR 3 | NP_940918.2 | |||
XM_006718224.2 | 2353 | UTR 3 | XP_006718287.1 | |||
XM_006718225.2 | 2353 | Intron | XP_006718288.1 | |||
XM_011520068.2 | 2353 | UTR 3 | XP_011518370.1 | |||
XM_011520069.2 | 2353 | Intron | XP_011518371.1 | |||
XM_011520070.2 | 2353 | Intron | XP_011518372.1 | |||
XM_011520071.2 | 2353 | Intron | XP_011518373.1 |