Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGACAATACTTACCTTTGTAGT[C/T]AGGTCCTGTTCTGCAGGAAGCGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA1 - proteasome subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143937.1 | 761 | Intron | NP_001137409.1 | |||
NM_002786.3 | 761 | Silent Mutation | CTA,CTG | L,L 205 | NP_002777.1 | |
NM_148976.2 | 761 | Silent Mutation | CTA,CTG | L,L 211 | NP_683877.1 |