Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTGTTCCTCCTGCCTCTCACCC[A/G]GCAGCCTTTGCCCTCTGGAGTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602631 MIM: 603240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A18 - solute carrier family 22 member 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001315501.1 | 1708 | Intron | NP_001302430.1 | |||
NM_001315502.1 | 1708 | Intron | NP_001302431.1 | |||
NM_002555.5 | 1708 | Intron | NP_002546.3 | |||
NM_183233.2 | 1708 | Intron | NP_899056.2 | |||
XM_011520141.2 | 1708 | Intron | XP_011518443.2 | |||
XM_011520142.2 | 1708 | Intron | XP_011518444.2 |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302862.1 | 1708 | UTR 5 | NP_001289791.1 | |||
NM_007105.3 | 1708 | UTR 5 | NP_009036.2 | |||
XM_017017832.1 | 1708 | UTR 5 | XP_016873321.1 | |||
XM_017017833.1 | 1708 | UTR 5 | XP_016873322.1 | |||
XM_017017834.1 | 1708 | UTR 5 | XP_016873323.1 | |||
XM_017017835.1 | 1708 | Intron | XP_016873324.1 | |||
XM_017017836.1 | 1708 | Intron | XP_016873325.1 |