Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGAGCAGCTAATTGAGAAGCTA[A/C]TTGAGGATGATGAAAACTCCCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150000 MIM: 150150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDHA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDHA - lactate dehydrogenase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LDHC - lactate dehydrogenase C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002301.4 | 211 | Missense Mutation | ATT,CTT | I,L 13 | NP_002292.1 | |
NM_017448.3 | 211 | Missense Mutation | ATT,CTT | I,L 13 | NP_059144.1 | |
XM_017017721.1 | 211 | UTR 5 | XP_016873210.1 | |||
XM_017017722.1 | 211 | UTR 5 | XP_016873211.1 |