Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGACTGAGCAAGTTTTAATGAC[A/G]TTGTTACATTCTTGACACTTGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF195 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF195 - zinc finger protein 195 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130519.2 | 1440 | Silent Mutation | AAC,AAT | N,N 284 | NP_001123991.1 | |
NM_001130520.2 | 1440 | Silent Mutation | AAC,AAT | N,N 307 | NP_001123992.1 | |
NM_001242841.1 | 1440 | Silent Mutation | AAC,AAT | N,N 288 | NP_001229770.1 | |
NM_001242842.1 | 1440 | Silent Mutation | AAC,AAT | N,N 262 | NP_001229771.1 | |
NM_001242843.1 | 1440 | Silent Mutation | AAC,AAT | N,N 239 | NP_001229772.1 | |
NM_001256823.1 | 1440 | UTR 3 | NP_001243752.1 | |||
NM_001256824.1 | 1440 | Silent Mutation | AAC,AAT | N,N 270 | NP_001243753.1 | |
NM_001256825.1 | 1440 | Silent Mutation | AAC,AAT | N,N 239 | NP_001243754.1 | |
NM_007152.4 | 1440 | Silent Mutation | AAC,AAT | N,N 235 | NP_009083.2 | |
XM_006718306.1 | 1440 | Silent Mutation | AAC,AAT | N,N 250 | XP_006718369.1 | |
XM_011520350.2 | 1440 | Silent Mutation | AAC,AAT | N,N 311 | XP_011518652.1 | |
XM_011520351.2 | 1440 | Silent Mutation | AAC,AAT | N,N 311 | XP_011518653.1 | |
XM_011520352.2 | 1440 | Silent Mutation | AAC,AAT | N,N 273 | XP_011518654.1 | |
XM_011520354.2 | 1440 | Intron | XP_011518656.2 | |||
XM_017018260.1 | 1440 | Silent Mutation | AAC,AAT | N,N 322 | XP_016873749.1 | |
XM_017018261.1 | 1440 | Silent Mutation | AAC,AAT | N,N 311 | XP_016873750.1 | |
XM_017018262.1 | 1440 | Silent Mutation | AAC,AAT | N,N 299 | XP_016873751.1 | |
XM_017018263.1 | 1440 | Silent Mutation | AAC,AAT | N,N 288 | XP_016873752.1 | |
XM_017018264.1 | 1440 | Silent Mutation | AAC,AAT | N,N 167 | XP_016873753.1 | |
XM_017018265.1 | 1440 | Silent Mutation | AAC,AAT | N,N 208 | XP_016873754.1 | |
XM_017018266.1 | 1440 | Silent Mutation | AAC,AAT | N,N 208 | XP_016873755.1 | |
XM_017018267.1 | 1440 | Silent Mutation | AAC,AAT | N,N 231 | XP_016873756.1 |