Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACTGATATGGCATTTGGCACA[C/T]AGTAGGCATTCAACAAATGAATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605493 | ||||||||||||||||||||
Literature Links: |
TRIM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM3 - tripartite motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248006.1 | Intron | NP_001234935.1 | ||||
NM_001248007.1 | Intron | NP_001234936.1 | ||||
NM_006458.3 | Intron | NP_006449.2 | ||||
NM_033278.3 | Intron | NP_150594.2 | ||||
XM_011519844.1 | Intron | XP_011518146.1 | ||||
XM_011519845.2 | Intron | XP_011518147.1 | ||||
XM_017017098.1 | Intron | XP_016872587.1 | ||||
XM_017017099.1 | Intron | XP_016872588.1 |