Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGGTTTTCCTCCTACCTTACCG[G/T]TTACTTCTCTGAGGCTTCTTTGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCUBE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCUBE2 - signal peptide, CUB domain and EGF like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170690.1 | 4298 | Intron | NP_001164161.1 | |||
NM_020974.2 | 4298 | Intron | NP_066025.2 | |||
XM_005253032.4 | 4298 | Intron | XP_005253089.2 | |||
XM_005253033.4 | 4298 | Intron | XP_005253090.2 | |||
XM_005253034.4 | 4298 | Intron | XP_005253091.2 | |||
XM_005253035.4 | 4298 | UTR 3 | XP_005253092.2 | |||
XM_005253036.4 | 4298 | Intron | XP_005253093.2 | |||
XM_005253037.4 | 4298 | Intron | XP_005253094.2 | |||
XM_011520246.2 | 4298 | Intron | XP_011518548.1 | |||
XM_011520247.2 | 4298 | Intron | XP_011518549.1 | |||
XM_011520248.2 | 4298 | Intron | XP_011518550.1 | |||
XM_017018080.1 | 4298 | Intron | XP_016873569.1 | |||
XM_017018081.1 | 4298 | Intron | XP_016873570.1 | |||
XM_017018082.1 | 4298 | UTR 3 | XP_016873571.1 |