Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCAGGTTTTATTGGCATCTTAA[C/T]CATTTTAAGTTAGAATTTTTGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNM1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNM1L - dynamin 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278463.1 | Intron | NP_001265392.1 | ||||
NM_001278464.1 | Intron | NP_001265393.1 | ||||
NM_001278465.1 | Intron | NP_001265394.1 | ||||
NM_001278466.1 | Intron | NP_001265395.1 | ||||
NM_005690.4 | Intron | NP_005681.2 | ||||
NM_012062.4 | Intron | NP_036192.2 | ||||
NM_012063.3 | Intron | NP_036193.2 | ||||
XM_005253282.4 | Intron | XP_005253339.1 | ||||
XM_011520543.2 | Intron | XP_011518845.1 | ||||
XM_011520544.2 | Intron | XP_011518846.1 | ||||
XM_017018663.1 | Intron | XP_016874152.1 | ||||
XM_017018664.1 | Intron | XP_016874153.1 | ||||
XM_017018665.1 | Intron | XP_016874154.1 |