Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAAAGAGAGGTCATGACAAAAC[C/T]ATCCCTGGCTTCCTTTGCTTCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN2B - glutamate ionotropic receptor NMDA type subunit 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000834.3 | 26712 | Intron | NP_000825.2 | |||
XM_005253351.3 | 26712 | UTR 3 | XP_005253408.1 | |||
XM_011520628.2 | 26712 | UTR 3 | XP_011518930.1 | |||
XM_011520629.2 | 26712 | UTR 3 | XP_011518931.1 | |||
XM_017019219.1 | 26712 | UTR 3 | XP_016874708.1 |