Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTTGCTCTAACAAAAGGGTAC[A/G]CTCAAGCTCCGCATAATCCCATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616359 MIM: 615998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ5 - coenzyme Q5, methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014868.4 | Intron | NP_055683.3 | ||||
XM_005254013.2 | Intron | XP_005254070.1 | ||||
XM_006719717.2 | Intron | XP_006719780.1 | ||||
XM_017020282.1 | Intron | XP_016875771.1 | ||||
XM_017020283.1 | Intron | XP_016875772.1 |