Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGCCAGTAATAGTGTCTGGAT[C/G]GGTCAGGAGCACGGCCTCTGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFC5 - replication factor C subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130112.2 | 1889 | Intron | NP_001123584.1 | |||
NM_001206801.1 | 1889 | Intron | NP_001193730.1 | |||
NM_007370.5 | 1889 | Intron | NP_031396.1 | |||
NM_181578.3 | 1889 | Intron | NP_853556.2 | |||
XM_011538643.2 | 1889 | Intron | XP_011536945.1 | |||
XM_011538645.2 | 1889 | Intron | XP_011536947.1 | |||
XM_017019779.1 | 1889 | Intron | XP_016875268.1 |
WSB2 - WD repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278557.1 | 1889 | UTR 3 | NP_001265486.1 | |||
NM_001278558.1 | 1889 | UTR 3 | NP_001265487.1 | |||
NM_018639.4 | 1889 | UTR 3 | NP_061109.1 | |||
XM_017019642.1 | 1889 | UTR 3 | XP_016875131.1 |