Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGACAGAAGAAGGAAGAGAGAC[A/G]GTAACAAGGAAAGAAGGATGGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607408 MIM: 607415 | ||||||||||||||||||||
Literature Links: |
DAOA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAOA - D-amino acid oxidase activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161812.1 | 281 | UTR 5 | NP_001155284.1 | |||
NM_001161814.1 | 281 | UTR 5 | NP_001155286.1 | |||
NM_172370.4 | 281 | Silent Mutation | ACA,ACG | T,T 54 | NP_758958.3 | |
XM_005254042.1 | 281 | Silent Mutation | ACA,ACG | T,T 54 | XP_005254099.1 |
DAOA-AS1 - DAOA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |