Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCACAGAGGCATTTACAGAGATC[A/T]TTATGTCCTTGGGTCTCTTACCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607408 MIM: 607415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAOA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAOA - D-amino acid oxidase activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161812.1 | 359 | Missense Mutation | CAT,CTT | H,L 16 | NP_001155284.1 | |
NM_001161814.1 | 359 | Silent Mutation | TCA,TCT | S,S 9 | NP_001155286.1 | |
NM_172370.4 | 359 | Silent Mutation | TCA,TCT | S,S 80 | NP_758958.3 | |
XM_005254042.1 | 359 | Silent Mutation | TCA,TCT | S,S 80 | XP_005254099.1 |
DAOA-AS1 - DAOA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |