Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGTACTATTTTCTAACAAGAAG[A/C]CTTCAGGGGAAATAATGGATCCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC4 - ATP binding cassette subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105515.2 | 4638 | Intron | NP_001098985.1 | |||
NM_001301829.1 | 4638 | UTR 3 | NP_001288758.1 | |||
NM_001301830.1 | 4638 | Intron | NP_001288759.1 | |||
NM_005845.4 | 4638 | UTR 3 | NP_005836.2 | |||
XM_005254025.2 | 4638 | UTR 3 | XP_005254082.1 | |||
XM_017020319.1 | 4638 | UTR 3 | XP_016875808.1 | |||
XM_017020320.1 | 4638 | Intron | XP_016875809.1 | |||
XM_017020321.1 | 4638 | UTR 3 | XP_016875810.1 | |||
XM_017020322.1 | 4638 | Intron | XP_016875811.1 |