Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGACTTCCTGTTACCAGACCA[C/T]GAAAACAGAATCAACTCTTACAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYSMD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYSMD2 - LysM domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMOD2 - tropomodulin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142885.1 | Intron | NP_001136357.1 | ||||
NM_014548.3 | Intron | NP_055363.1 | ||||
XM_017022089.1 | Intron | XP_016877578.1 |