Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGTCTATAACACCGCTTTTATA[G/T]AACTGAGCAATCAAAATGTATGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR72 - WD repeat domain 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277176.1 | 3713 | UTR 3 | NP_001264105.1 | |||
NM_182758.3 | 3713 | UTR 3 | NP_877435.3 | |||
XM_011521433.2 | 3713 | Intron | XP_011519735.1 | |||
XM_011521435.2 | 3713 | Intron | XP_011519737.1 | |||
XM_011521436.2 | 3713 | Intron | XP_011519738.1 | |||
XM_011521437.2 | 3713 | Intron | XP_011519739.1 | |||
XM_017022061.1 | 3713 | Intron | XP_016877550.1 |