Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATAAAAAAAAATAAAATCTGGA[C/T]GTAAAATGTTAATTTATAGTACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTFMT - mitochondrial methionyl-tRNA formyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139242.3 | 2503 | UTR 3 | NP_640335.2 | |||
XM_005254158.4 | 2503 | UTR 3 | XP_005254215.2 |