Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTTTGAGCGACCCCTCTCCCC[C/T]GGGCCTGGCTGCGCGCTGCTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2Q2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2Q2 - ubiquitin conjugating enzyme E2 Q2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145335.1 | 384 | Missense Mutation | CCG,CTG | P,L 25 | NP_001138807.1 | |
NM_001284382.1 | 384 | Intron | NP_001271311.1 | |||
NM_173469.3 | 384 | Intron | NP_775740.1 | |||
XM_005254787.1 | 384 | Intron | XP_005254844.1 | |||
XM_005254788.1 | 384 | Intron | XP_005254845.1 | |||
XM_006720770.1 | 384 | Intron | XP_006720833.1 | |||
XM_011522228.1 | 384 | Intron | XP_011520530.1 | |||
XM_017022726.1 | 384 | Missense Mutation | CCG,CTG | P,L 25 | XP_016878215.1 | |
XM_017022727.1 | 384 | Intron | XP_016878216.1 | |||
XM_017022728.1 | 384 | Intron | XP_016878217.1 |