Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAAATACCTGTTGGTGTCATCC[C/T]TGTTTGAGCCAGGGTCTGAGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610894 | ||||||||||||||||||||
Literature Links: |
NRG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRG4 - neuregulin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138573.3 | Intron | NP_612640.1 |
TMEM266 - transmembrane protein 266 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152335.2 | Intron | NP_689548.2 | ||||
XM_005254160.2 | Intron | XP_005254217.1 | ||||
XM_017021915.1 | Intron | XP_016877404.1 | ||||
XM_017021916.1 | Intron | XP_016877405.1 | ||||
XM_017021917.1 | Intron | XP_016877406.1 |