Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGTCATCATAACTGAAGAGAGA[C/T]AAATCCAGATAGGGGCTACCGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DET1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DET1 - de-etiolated homolog 1 (Arabidopsis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144074.2 | 1595 | Silent Mutation | TTA,TTG | L,L 463 | NP_001137546.1 | |
NM_001321594.1 | 1595 | Silent Mutation | TTA,TTG | L,L 429 | NP_001308523.1 | |
NM_001321596.1 | 1595 | Silent Mutation | TTA,TTG | L,L 463 | NP_001308525.1 | |
NM_017996.4 | 1595 | Silent Mutation | TTA,TTG | L,L 474 | NP_060466.2 |