Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTGGAAAAGATGTAGAAATCAC[A/C]TTGGCCCATCCCCCCTCATTTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617029 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEMA4B - semaphorin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324029.1 | Intron | NP_001310958.1 | ||||
NM_001324030.1 | Intron | NP_001310959.1 | ||||
NM_001324031.1 | Intron | NP_001310960.1 | ||||
NM_001324032.1 | Intron | NP_001310961.1 | ||||
NM_001324033.1 | Intron | NP_001310962.1 | ||||
NM_001324034.1 | Intron | NP_001310963.1 | ||||
NM_020210.3 | Intron | NP_064595.2 | ||||
NM_198925.2 | Intron | NP_945119.1 |