Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATTAGCCCCCCAGCCCTCATG[A/G]ACTGAAAAAGAGAGACAGATGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
VWA3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VWA3A - von Willebrand factor A domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173615.4 | Intron | NP_775886.3 | ||||
XM_011545742.2 | Intron | XP_011544044.1 | ||||
XM_011545744.2 | Intron | XP_011544046.1 | ||||
XM_011545745.1 | Intron | XP_011544047.1 | ||||
XM_011545746.2 | Intron | XP_011544048.1 | ||||
XM_017022947.1 | Intron | XP_016878436.1 | ||||
XM_017022948.1 | Intron | XP_016878437.1 | ||||
XM_017022949.1 | Intron | XP_016878438.1 |