Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCGGTTCTCCCTCTCCACTGA[A/G]GACAGAATTTGAGGAAATGAGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF40 - ring finger protein 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207033.1 | 4000 | Intron | NP_001193962.1 | |||
NM_001207034.1 | 4000 | Intron | NP_001193963.1 | |||
NM_001286572.2 | 4000 | Intron | NP_001273501.1 | |||
NM_014771.3 | 4000 | Intron | NP_055586.1 | |||
XM_011545997.1 | 4000 | Intron | XP_011544299.1 |
ZNF629 - zinc finger protein 629 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080417.1 | 4000 | UTR 3 | NP_001073886.1 | |||
XM_005255222.3 | 4000 | UTR 3 | XP_005255279.1 |