Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTACCAGTGGTTATCATTTTA[A/G]TTCTTCCATTTAGTTCTATGAGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARN - poly(A)-specific ribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134477.2 | Intron | NP_001127949.1 | ||||
NM_001242992.1 | Intron | NP_001229921.1 | ||||
NM_002582.3 | Intron | NP_002573.1 | ||||
XM_011522510.2 | Intron | XP_011520812.1 | ||||
XM_011522511.2 | Intron | XP_011520813.1 | ||||
XM_011522513.2 | Intron | XP_011520815.1 | ||||
XM_011522514.1 | Intron | XP_011520816.1 | ||||
XM_017023258.1 | Intron | XP_016878747.1 | ||||
XM_017023259.1 | Intron | XP_016878748.1 | ||||
XM_017023260.1 | Intron | XP_016878749.1 |