Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTGTTGCCCTGCTGAAAAACCT[A/G]TGTACCAGTTGGTCCCTTTGGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP93 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP93 - nucleoporin 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242795.1 | Intron | NP_001229724.1 | ||||
NM_001242796.1 | Intron | NP_001229725.1 | ||||
NM_014669.4 | Intron | NP_055484.3 | ||||
XM_005256263.3 | Intron | XP_005256320.1 |