Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGGACTTCCTGCCCCTAGAGAC[A/G]ACCAACAACAACAACAACCACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPD5 - PAP associated domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040284.2 | 370 | Silent Mutation | ACA,ACG | T,T 93 | NP_001035374.2 | |
NM_001040285.2 | 370 | Silent Mutation | ACA,ACG | T,T 93 | NP_001035375.2 | |
XM_005256092.4 | 370 | Silent Mutation | ACA,ACG | T,T 108 | XP_005256149.1 | |
XM_005256093.4 | 370 | Silent Mutation | ACA,ACG | T,T 94 | XP_005256150.2 | |
XM_011523275.2 | 370 | Silent Mutation | ACA,ACG | T,T 94 | XP_011521577.1 | |
XM_011523276.2 | 370 | Silent Mutation | ACA,ACG | T,T 94 | XP_011521578.1 | |
XM_011523278.2 | 370 | Intron | XP_011521580.1 | |||
XM_017023555.1 | 370 | Silent Mutation | ACA,ACG | T,T 88 | XP_016879044.1 |