Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGACCTCCCTTTAAAAAAGAAAG[C/T]GAGGGACACATCAGTGTGTAAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371374 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371374 - uncharacterized LOC105371374 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLDC1 - TBC/LysM-associated domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020947.3 | 4748 | UTR 3 | NP_065998.3 | |||
XM_005256074.3 | 4748 | UTR 3 | XP_005256131.1 | |||
XM_005256075.2 | 4748 | Intron | XP_005256132.1 | |||
XM_006721240.2 | 4748 | Intron | XP_006721303.1 | |||
XM_011523248.2 | 4748 | Intron | XP_011521550.1 | |||
XM_011523249.2 | 4748 | Intron | XP_011521551.1 | |||
XM_017023510.1 | 4748 | Intron | XP_016878999.1 | |||
XM_017023511.1 | 4748 | Intron | XP_016879000.1 | |||
XM_017023512.1 | 4748 | Intron | XP_016879001.1 |