Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAACAGTGACATCAAAGACTTT[A/G]CGTGTCTCTTCTTGACCACTCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164010 MIM: 615409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP1A - charged multivesicular body protein 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA33 - spermatogenesis associated 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271907.1 | Intron | NP_001258836.1 | ||||
NM_001271908.1 | Intron | NP_001258837.1 | ||||
NM_001271909.1 | Intron | NP_001258838.1 | ||||
NM_001271910.1 | Intron | NP_001258839.1 | ||||
NM_153025.2 | Intron | NP_694570.1 | ||||
XM_006721133.2 | Intron | XP_006721196.1 |