Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAACAAAGCCACTATTTGCATAT[C/T]TGGGAAAGGAAACATATTGCTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612543 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP36 - ubiquitin specific peptidase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321291.1 | 4508 | UTR 3 | NP_001308220.1 | |||
XM_005257542.2 | 4508 | UTR 3 | XP_005257599.1 | |||
XM_005257543.2 | 4508 | UTR 3 | XP_005257600.1 | |||
XM_011525065.1 | 4508 | UTR 3 | XP_011523367.1 | |||
XM_011525066.2 | 4508 | UTR 3 | XP_011523368.1 | |||
XM_011525067.2 | 4508 | Intron | XP_011523369.1 | |||
XM_011525068.1 | 4508 | UTR 3 | XP_011523370.1 | |||
XM_011525069.2 | 4508 | UTR 3 | XP_011523371.1 | |||
XM_011525070.1 | 4508 | UTR 3 | XP_011523372.1 | |||
XM_011525071.2 | 4508 | Intron | XP_011523373.1 | |||
XM_017024900.1 | 4508 | Intron | XP_016880389.1 | |||
XM_017024901.1 | 4508 | UTR 3 | XP_016880390.1 |