Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGTATGAAAAAAACTTTAAAA[G/T]TCAGCAAGCAATTCTTAATGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600849 MIM: 613814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC19 - tetratricopeptide repeat domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271420.1 | 9953 | Intron | NP_001258349.1 | |||
NM_017775.3 | 9953 | Intron | NP_060245.3 | |||
XM_017024801.1 | 9953 | Intron | XP_016880290.1 | |||
XM_017024802.1 | 9953 | Intron | XP_016880291.1 |