Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTTACCTAGGAAAGTTCCAGC[G/A]AACCCCAGTGCAAAGAAGCTGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602391 | ||||||||||||||||||||
Literature Links: |
PEMT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEMT - phosphatidylethanolamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267551.1 | 314 | Silent Mutation | NP_001254480.1 | |||
NM_001267552.1 | 314 | Silent Mutation | NP_001254481.1 | |||
NM_007169.2 | 314 | Silent Mutation | NP_009100.2 | |||
NM_148172.2 | 314 | Silent Mutation | NP_680477.1 | |||
NM_148173.1 | 314 | Silent Mutation | NP_680478.1 | |||
XM_006721418.3 | 314 | Silent Mutation | XP_006721481.2 | |||
XM_017024016.1 | 314 | Silent Mutation | XP_016879505.1 |