Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGAAATTGAGAAAACAGTGTA[A/G]GCAAAATCAGTAGTAAAATAAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC728392 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC728392 - uncharacterized LOC728392 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162371.2 | 5016 | Intron | NP_001155843.1 |
NLRP1 - NLR family pyrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033053.2 | 5016 | UTR 3 | NP_001028225.1 | |||
NM_014922.4 | 5016 | Intron | NP_055737.1 | |||
NM_033004.3 | 5016 | Intron | NP_127497.1 | |||
NM_033006.3 | 5016 | Intron | NP_127499.1 | |||
NM_033007.3 | 5016 | Intron | NP_127500.1 |