Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGGGGGCGGGGGGCGTTGTCAG[A/G]CAGGGCTACTAGGCTGCAAAGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610684 MIM: 602151 MIM: 605125 MIM: 612910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTDNEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTDNEP1 - CTD nuclear envelope phosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DVL2 - dishevelled segment polarity protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GABARAP - GABA type A receptor-associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF23 - PHD finger protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284517.1 | Intron | NP_001271446.1 | ||||
NM_001284518.1 | Intron | NP_001271447.1 | ||||
NM_024297.2 | Intron | NP_077273.2 |