Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGGAGAATCACACACAGCTTC[C/T]GCTGGATGGCCCAGGTGTTGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ODF4 - outer dense fiber of sperm tails 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319953.1 | 411 | Intron | NP_001306882.1 | |||
NM_153007.4 | 411 | Missense Mutation | CGC,TGC | R,C 77 | NP_694552.2 | |
XM_005256476.3 | 411 | Intron | XP_005256533.1 | |||
XM_011523688.1 | 411 | Intron | XP_011521990.1 |