Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTCAGAGCAGACAGAGCCACA[A/G]CAGGAGCTGACCATGGCGTCAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRTAP4-2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRTAP4-2 - keratin associated protein 4-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP4-3 - keratin associated protein 4-3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033187.1 | 15 | Silent Mutation | TGC,TGT | C,C 5 | NP_149443.1 |
KRTAP4-4 - keratin associated protein 4-4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |