Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGATCATTCAAAGTTCAAGATG[A/G]GCCAATGGATTTTCACATAATAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616317 MIM: 613175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDPD1 - glycerophosphodiester phosphodiesterase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165993.1 | Intron | NP_001159465.1 | ||||
NM_001165994.1 | Intron | NP_001159466.1 | ||||
NM_182569.3 | Intron | NP_872375.2 | ||||
XM_017024521.1 | Intron | XP_016880010.1 |
SMG8 - SMG8, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |