Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGACCTACAGCTGACCCAGTGA[C/T]CTCAGATGCTGCCAAGTCCTGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604067 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS9 - regulator of G-protein signaling 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081955.2 | Intron | NP_001075424.1 | ||||
NM_001165933.1 | Intron | NP_001159405.1 | ||||
NM_003835.3 | Intron | NP_003826.2 | ||||
XM_011525426.2 | Intron | XP_011523728.1 |