Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCTTTTGAAGCATTTACTTTC[C/T]GAGTCGCCAGCTGGTTCCAAAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEOX1 - mesenchyme homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040002.1 | 1042 | UTR 3 | NP_001035091.1 | |||
NM_004527.3 | 1042 | UTR 3 | NP_004518.1 | |||
NM_013999.3 | 1042 | UTR 3 | NP_054705.1 | |||
XM_011524818.1 | 1042 | Intron | XP_011523120.1 |