Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAATGAAGGGCAGGGTGTCCATC[C/T]GTGACAGCCGCCAGGAGCTCTCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD300LG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD300LG - CD300 molecule like family member g | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168322.1 | 684 | Missense Mutation | CGT,TGT | R,C 82 | NP_001161794.1 | |
NM_001168323.1 | 684 | Missense Mutation | CGT,TGT | R,C 82 | NP_001161795.1 | |
NM_001168324.1 | 684 | Missense Mutation | CGT,TGT | R,C 82 | NP_001161796.1 | |
NM_145273.3 | 684 | Missense Mutation | CGT,TGT | R,C 82 | NP_660316.2 | |
XM_005257075.2 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_005257132.1 | |
XM_005257076.2 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_005257133.1 | |
XM_006721721.2 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_006721784.1 | |
XM_006721722.2 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_006721785.1 | |
XM_011524384.2 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_011522686.1 | |
XM_017024235.1 | 684 | Missense Mutation | CGT,TGT | R,C 82 | XP_016879724.1 |