Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGACACACAGGGGATATGGCTG[C/G]TTTTTTTTTTTTTTTTTACTTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613907 | ||||||||||||||||||||
Literature Links: |
MIR6129 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6129 - microRNA 6129 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF652 - zinc finger protein 652 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145365.1 | 11339 | UTR 3 | NP_001138837.1 | |||
NM_014897.2 | 11339 | UTR 3 | NP_055712.1 | |||
XM_005257166.2 | 11339 | UTR 3 | XP_005257223.1 |