Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCATGAAAGTAAATAGAAAAA[A/T]AATCACAGTTCAAGAATAACATAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614953 | ||||||||||||||||||||
Literature Links: |
SLFN11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLFN11 - schlafen family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104587.1 | 3952 | Intron | NP_001098057.1 | |||
NM_001104588.1 | 3952 | Intron | NP_001098058.1 | |||
NM_001104589.1 | 3952 | Intron | NP_001098059.1 | |||
NM_001104590.1 | 3952 | Intron | NP_001098060.1 | |||
NM_152270.3 | 3952 | Intron | NP_689483.3 | |||
XM_005258068.3 | 3952 | UTR 3 | XP_005258125.1 | |||
XM_006722171.3 | 3952 | UTR 3 | XP_006722234.1 | |||
XM_011525465.2 | 3952 | UTR 3 | XP_011523767.1 | |||
XM_017025299.1 | 3952 | UTR 3 | XP_016880788.1 |