Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTTTCAACTCCTTTTCCTCCT[C/T]CCCTTCCTCTTGAAACATCTCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611398 MIM: 608417 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf50 - chromosome 17 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145272.3 | Intron | NP_660315.2 |
GAS2L2 - growth arrest specific 2 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP28 - matrix metallopeptidase 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032278.2 | Intron | NP_001027449.1 | ||||
NM_024302.4 | Intron | NP_077278.1 | ||||
NM_032950.3 | Intron | NP_116568.1 | ||||
XM_011525225.1 | Intron | XP_011523527.1 | ||||
XM_011525226.2 | Intron | XP_011523528.1 | ||||
XM_011525227.1 | Intron | XP_011523529.1 | ||||
XM_011525228.1 | Intron | XP_011523530.1 | ||||
XM_011525229.2 | Intron | XP_011523531.1 | ||||
XM_011525230.1 | Intron | XP_011523532.1 | ||||
XM_011525231.1 | Intron | XP_011523533.1 | ||||
XM_011525232.2 | Intron | XP_011523534.2 | ||||
XM_017025061.1 | Intron | XP_016880550.1 | ||||
XM_017025062.1 | Intron | XP_016880551.1 | ||||
XM_017025063.1 | Intron | XP_016880552.1 | ||||
XM_017025064.1 | Intron | XP_016880553.1 |