Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTTTTAGAATAATGGTTCTAA[A/G]TGTTTTTTACAACACTGTATGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL4 - methyltransferase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308401.1 | 2189 | UTR 3 | NP_001295330.1 | |||
NM_022840.4 | 2189 | UTR 3 | NP_073751.3 | |||
XM_005258132.3 | 2189 | UTR 3 | XP_005258189.1 | |||
XM_005258133.2 | 2189 | UTR 3 | XP_005258190.1 | |||
XM_011525730.2 | 2189 | UTR 3 | XP_011524032.1 |