Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTCCGCTTGCGCTTGGGTAT[C/G]TGGCTTGGGGGGCGGAGTGGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM200C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM200C - transmembrane protein 200C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080209.1 | 110 | Missense Mutation | CAC,CAG | H,Q 23 | NP_001073678.1 | |
XM_005258130.4 | 110 | Missense Mutation | CAC,CAG | H,Q 23 | XP_005258187.1 | |
XM_011525729.2 | 110 | Missense Mutation | CAC,CAG | H,Q 23 | XP_011524031.1 |