Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACTATTGCAATAAAAATTACAT[A/G]TAACAAGTTTGATTCTTTGCTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610432 MIM: 616319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF125 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF125 - ring finger protein 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF138 - ring finger protein 138 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191324.1 | Intron | NP_001178253.2 | ||||
NM_016271.4 | Intron | NP_057355.2 | ||||
NM_198128.2 | Intron | NP_937761.1 | ||||
XM_005258285.1 | Intron | XP_005258342.1 | ||||
XM_005258286.1 | Intron | XP_005258343.1 |