Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACATTGACTCTTCTGAATACTAC[A/G]TGAAATCTAATCTAAAAAGGTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBBP8 - RB binding protein 8, endonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002894.2 | Intron | NP_002885.1 | ||||
NM_203291.1 | Intron | NP_976036.1 | ||||
NM_203292.1 | Intron | NP_976037.1 | ||||
XM_005258325.2 | Intron | XP_005258382.1 | ||||
XM_005258326.3 | Intron | XP_005258383.1 | ||||
XM_006722519.2 | Intron | XP_006722582.1 | ||||
XM_006722520.2 | Intron | XP_006722583.1 | ||||
XM_006722521.2 | Intron | XP_006722584.1 | ||||
XM_011526132.2 | Intron | XP_011524434.1 | ||||
XM_017025915.1 | Intron | XP_016881404.1 | ||||
XM_017025916.1 | Intron | XP_016881405.1 |