Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCACACCTGCTTCCACTCTGAC[C/T]TCCACATCTAACTCATGTAATTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF607 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF607 - zinc finger protein 607 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172677.1 | 3149 | UTR 3 | NP_001166148.1 | |||
NM_032689.4 | 3149 | UTR 3 | NP_116078.4 | |||
XM_006723435.3 | 3149 | UTR 3 | XP_006723498.1 | |||
XM_006723436.2 | 3149 | UTR 3 | XP_006723499.1 | |||
XM_006723437.2 | 3149 | UTR 3 | XP_006723500.1 | |||
XM_006723440.1 | 3149 | UTR 3 | XP_006723503.1 | |||
XM_011527414.2 | 3149 | UTR 3 | XP_011525716.1 |
ZNF781 - zinc finger protein 781 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |