Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGCCATTGAGGAAGGCTTCTCT[G/T]GCTTCTCCCTGTTCATCCAAGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609744 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CADM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CADM4 - cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145296.1 | 1065 | Silent Mutation | GCA,GCC | A,A 369 | NP_660339.1 | |
XM_005258620.2 | 1065 | Silent Mutation | GCA,GCC | A,A 326 | XP_005258677.1 | |
XM_017026452.1 | 1065 | Silent Mutation | GCA,GCC | A,A 350 | XP_016881941.1 |
SRRM5 - serine/arginine repetitive matrix 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF428 - zinc finger protein 428 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |