Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGGAAGTGTAATCGAGGAGCA[C/T]CATGTAGTCGCAACACTTGAGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF223 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF223 - zinc finger protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013361.5 | 320 | Intron | NP_037493.3 | |||
XM_017027258.1 | 320 | UTR 5 | XP_016882747.1 | |||
XM_017027259.1 | 320 | Intron | XP_016882748.1 | |||
XM_017027260.1 | 320 | Intron | XP_016882749.1 |